Learn More
REPLIGEN CORPORATION PharmaPure Tubing, Low Particulate Spallation, Use In Vaccine Manufacturing, 25 ft
SDP
Catalog No. NC2955609
Encompass
PharmaPure Peristaltic Pump Tubing is designed for critical fluid transfer in pharmaceuticals, offering superior flex life and low particulate spallation. Its robust construction extends pump life and ensure product integrity, making it ideal for media processing and aseptic filling. This tubing withstands autoclaving and meets stringent regulatory standards, ensuring reliable and efficient operation.
Catalog No. NC2955609 Supplier REPLIGEN CORPORATION Supplier No. ACTUP1625N
PharmaPure Peristaltic Pump Tubing is designed for critical fluid transfer in pharmaceuticals, offering superior flex life and low particulate spallation. Its robust construction extends pump life and ensure product integrity, making it ideal for media processing and aseptic filling. This tubing withstands autoclaving and meets stringent regulatory standards, ensuring reliable and efficient operation.