Learn More
Accuflow Systems Saint-Gobain Tubing, PharMed BPT, 3/16 in ID, 25 ft
Catalog No. 502365772
Encompass_Preferred
Saint-Gobain PharMed BPT tubing is a high-performance peristaltic pump tubing formulated for biocompatible fluid transfer in pharmaceutical, biotechnology, and diagnostic processes. It is engineered to withstand repeated peristaltic pumping, minimize particulate generation, and protect shear-sensitive fluids during transfer and processing.
  • 3/16 in ID, 5/16 in OD, 1/16 in wall, 25 ft length.
  • Proprietary peristaltic-grade formulation, opaque cream appearance.
  • Meets USP <88> Class VI and ISO 10993-5 biocompatibility testing.
  • Maximum service temperature 275°F (135°C); low temperature embrittlement -75°F (-59°C).
  • Very good general chemical resistance; excellent resistance to acids, alkalis, and oxidizers.
  • Compatible with repeated autoclaving and gamma irradiation up to 50 kGy.
  • Suitable for cell harvest, bioreactor process lines, aseptic filling, and shear-sensitive fluid transfer.
Catalog No. 50-236-5772 Supplier Accuflow Systems Supplier No. AY242012
Saint-Gobain PharMed BPT tubing is a high-performance peristaltic pump tubing formulated for biocompatible fluid transfer in pharmaceutical, biotechnology, and diagnostic processes. It is engineered to withstand repeated peristaltic pumping, minimize particulate generation, and protect shear-sensitive fluids during transfer and processing.
  • 3/16 in ID, 5/16 in OD, 1/16 in wall, 25 ft length.
  • Proprietary peristaltic-grade formulation, opaque cream appearance.
  • Meets USP <88> Class VI and ISO 10993-5 biocompatibility testing.
  • Maximum service temperature 275°F (135°C); low temperature embrittlement -75°F (-59°C).
  • Very good general chemical resistance; excellent resistance to acids, alkalis, and oxidizers.
  • Compatible with repeated autoclaving and gamma irradiation up to 50 kGy.
  • Suitable for cell harvest, bioreactor process lines, aseptic filling, and shear-sensitive fluid transfer.