Saint-Gobain PharMed BPT peristaltic pump tubing provides a biocompatible fluid-contact surface, extended pump life compared with silicone, and resistance to common sterilization methods suitable for bioprocessing and laboratory fluid transfer.
- Formulated thermoplastic elastomer for peristaltic pumping.
- Dimensions: 1/16 in ID, 1/8 in OD, 1/32 in wall, 25 ft length.
- Opaque cream color to protect light-sensitive fluids.
- Meets USP Class VI and ISO 10993-5 biocompatibility requirements.
- Sterilization: autoclavable (30 min at 121 °C), tolerant to 50 kGy gamma radiation.
- Maximum service temperature 275 °F (135 °C); low temperature embrittlement at -75 °F (-59 °C).
- Max working pressure 24 psi at 73 °F, 14 psi at 180 °F; vacuum rating 29.9 in Hg.
- Typical uses: cell harvest, bioreactor lines, aseptic filling, filtration, and diagnostics.