PharMed BPT tubing is a biocompatible, high-performance peristaltic pump tubing formulated for sensitive bioprocess and diagnostic applications. It delivers extended flex life and chemical resistance compared with standard silicone tubing, and supports common cleaning and sterilization methods.
- 1/8 in inner diameter, 1/4 in outer diameter, 25 ft length.
- Opaque cream appearance, Shore A 64 hardness.
- Meets USP Class VI, USP <87>, and ISO 10993-5 biocompatibility standards.
- Autoclavable for repeated cycles; compatible with gamma sterilization up to 50 kGy.
- Temperature range up to 275 °F (135 °C); low temperature embrittlement at -75 °F (-59 °C).
- Very good chemical resistance; excellent resistance to acids, alkalis, and oxidizers.
- Superior peristaltic pump performance versus silicone; demonstrated 1,000+ hours in comparative tests.
- Opaque to visible and UV light, protecting light-sensitive fluids.