Learn More
Accuflow Systems Saint-Gobain PharMed BPT Tubing, Biocompatible, 1/4 in ID, 25 ft
Catalog No. 502365912
Encompass_Preferred
Saint-Gobain PharMed BPT tubing is a biocompatible, high-flex-life peristaltic pump tubing formulated for reliable fluid transfer in bioprocessing, diagnostics, and laboratory applications. It offers improved chemical resistance and reduced gas permeability versus silicone, and is compatible with common sterilization methods for aseptic processing.
  • Designed for peristaltic pumping with superior flex life
  • Biocompatible; meets USP <88> Class VI and ISO 10993-5
  • 1/4 in inner diameter, 3/8 in outer diameter, 25 ft length
  • Opaque cream color protects light-sensitive fluids
  • Good chemical resistance to acids, alkalis, and oxidizers
  • Autoclavable and gamma radiation resistant (up to 50 kGy)
  • Wide temperature range; service to 275°F, embrittlement at -75°F
Catalog No. 50-236-5912 Supplier Accuflow Systems Supplier No. AY242017
Saint-Gobain PharMed BPT tubing is a biocompatible, high-flex-life peristaltic pump tubing formulated for reliable fluid transfer in bioprocessing, diagnostics, and laboratory applications. It offers improved chemical resistance and reduced gas permeability versus silicone, and is compatible with common sterilization methods for aseptic processing.
  • Designed for peristaltic pumping with superior flex life
  • Biocompatible; meets USP <88> Class VI and ISO 10993-5
  • 1/4 in inner diameter, 3/8 in outer diameter, 25 ft length
  • Opaque cream color protects light-sensitive fluids
  • Good chemical resistance to acids, alkalis, and oxidizers
  • Autoclavable and gamma radiation resistant (up to 50 kGy)
  • Wide temperature range; service to 275°F, embrittlement at -75°F