Learn More
Accuflow Systems Saint-Gobain PharMed BPT Tubing, 1/4 in ID, 1/2 in OD
Catalog No. 502365925
Encompass_Preferred
Saint-Gobain PharMed BPT tubing is a biocompatible thermoplastic tubing formulated for peristaltic pump applications in bioprocessing, diagnostics, and laboratory fluid handling. It offers chemical and oxidation resistance, low gas permeability, and durability under peristaltic pumping to maintain fluid integrity for shear-sensitive and sterile processes.
  • 1/4 in inner diameter, 1/2 in outer diameter, 1/8 in wall, 25 ft per package.
  • Opaque cream color for light sensitivity protection.
  • Formulated for peristaltic pump compatibility and extended pump life.
  • Meets USP <88> Class VI, USP <87>, and ISO 10993-5 biocompatibility requirements.
  • Suitable for cell culture, media transfer, aseptic filling, and diagnostics.
  • Low extractables with documented physiochemical and extractable testing.
Catalog No. 50-236-5925 Supplier Accuflow Systems Supplier No. AY242019
Saint-Gobain PharMed BPT tubing is a biocompatible thermoplastic tubing formulated for peristaltic pump applications in bioprocessing, diagnostics, and laboratory fluid handling. It offers chemical and oxidation resistance, low gas permeability, and durability under peristaltic pumping to maintain fluid integrity for shear-sensitive and sterile processes.
  • 1/4 in inner diameter, 1/2 in outer diameter, 1/8 in wall, 25 ft per package.
  • Opaque cream color for light sensitivity protection.
  • Formulated for peristaltic pump compatibility and extended pump life.
  • Meets USP <88> Class VI, USP <87>, and ISO 10993-5 biocompatibility requirements.
  • Suitable for cell culture, media transfer, aseptic filling, and diagnostics.
  • Low extractables with documented physiochemical and extractable testing.