Learn More
Accuflow Systems Saint-Gobain Peristaltic Tubing, PharMed BPT, 1/16 in ID, 25 ft
Catalog No. 502365928
Encompass_Preferred
PharMed BPT peristaltic tubing offers extended flex life, broad chemical resistance, and documented biocompatibility for reliable fluid transfer in bioprocess and laboratory systems. The tubing is formulated for peristaltic pump use, tolerates autoclaving and gamma sterilization, and provides an opaque barrier for light-sensitive media.
  • Designed for peristaltic pump applications with extended flex life and reduced downtime.
  • 1/16 in inner diameter, 3/16 in outer diameter, 1/16 in wall thickness, supplied in 25 ft lengths.
  • Opaque cream thermoplastic with Shore A hardness 64 and maximum service temperature 275 °F (135 °C).
  • Meets USP <88> Class VI, USP <87>, and ISO 10993-5 biocompatibility standards.
  • Withstands repeated autoclaving (up to five cycles) and gamma irradiation up to 50 kGy.
  • Good resistance to acids, alkalis, and oxidizers, and lower gas permeability than silicone tubing.
Catalog No. 50-236-5928 Supplier Accuflow Systems Supplier No. AY242003
PharMed BPT peristaltic tubing offers extended flex life, broad chemical resistance, and documented biocompatibility for reliable fluid transfer in bioprocess and laboratory systems. The tubing is formulated for peristaltic pump use, tolerates autoclaving and gamma sterilization, and provides an opaque barrier for light-sensitive media.
  • Designed for peristaltic pump applications with extended flex life and reduced downtime.
  • 1/16 in inner diameter, 3/16 in outer diameter, 1/16 in wall thickness, supplied in 25 ft lengths.
  • Opaque cream thermoplastic with Shore A hardness 64 and maximum service temperature 275 °F (135 °C).
  • Meets USP <88> Class VI, USP <87>, and ISO 10993-5 biocompatibility standards.
  • Withstands repeated autoclaving (up to five cycles) and gamma irradiation up to 50 kGy.
  • Good resistance to acids, alkalis, and oxidizers, and lower gas permeability than silicone tubing.