Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain PharMed BPT Tubing, 5/16 in ID, 25 ft

Catalog No. 502365929
Encompass_Preferred

PharMed BPT peristaltic pump tubing is a biocompatible thermoplastic engineered for long flex life and reliable low-shear fluid transfer. It is opaque to visible and UV light, offers broad resistance to acids, alkalis, and oxidizing agents, and is intended for medical, diagnostic, and laboratory applications requiring biocompatibility and chemical resistance.

  • Peristaltic pump tubing for low-shear fluid transfer.
  • Material: biocompatible thermoplastic (BPT formulation).
  • I.D. 5/16 in (7.9 mm), O.D. 7/16 in (11.1 mm), wall 1/16 in (1.6 mm).
  • Length: 25 ft (7.6 m) packaged.
  • Opaque to visible and UV light to protect light-sensitive fluids.
  • Resistant to acids, alkalis, and oxidizing agents.
  • Typical applications: diagnostic analyzers, reagent transfer, infusion systems, and sample transport.

Catalog No. 50-236-5929 Supplier Accuflow Systems Supplier No. AY242022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PharMed BPT peristaltic pump tubing is a biocompatible thermoplastic engineered for long flex life and reliable low-shear fluid transfer. It is opaque to visible and UV light, offers broad resistance to acids, alkalis, and oxidizing agents, and is intended for medical, diagnostic, and laboratory applications requiring biocompatibility and chemical resistance.

  • Peristaltic pump tubing for low-shear fluid transfer.
  • Material: biocompatible thermoplastic (BPT formulation).
  • I.D. 5/16 in (7.9 mm), O.D. 7/16 in (11.1 mm), wall 1/16 in (1.6 mm).
  • Length: 25 ft (7.6 m) packaged.
  • Opaque to visible and UV light to protect light-sensitive fluids.
  • Resistant to acids, alkalis, and oxidizing agents.
  • Typical applications: diagnostic analyzers, reagent transfer, infusion systems, and sample transport.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.