Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High-Pressure Pump Tubing, PharMed BPT, L/S 16HP; 25 ft
Catalog No. NC1270480
Encompass
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige High-Pressure PharMed BPT tubing is designed for use with the L/S High-Performance pump head models 77250-62 and 77250-82 only.
Please note that the maximum drive speed for these High-Pressure PharMed BPT tubing is 100 RPM.
Catalog No. NC1270480 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0566416
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige High-Pressure PharMed BPT tubing is designed for use with the L/S High-Performance pump head models 77250-62 and 77250-82 only.
Please note that the maximum drive speed for these High-Pressure PharMed BPT tubing is 100 RPM.