Learn More
AVANTOR MASTERFLEX (VWR LLC) PharMed BPT High-Pressure Tubing, L/S 24HP, 25'
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0566424
Encompass
Masterflex PharMed BPT High-Pressure Tubing, L/S 24HP, 25', Designed to pump at pressures up to 150 psig (10.2 bar)Available in sizes 14, 16, 15, and 24,With size 14, one can pump continusously at 150 PSIG.Has USP Class VI regulatory approval
Catalog No. NC1270482
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.