Learn More
Accuflow Systems Saint-Gobain PharMed BPT Tubing, 1/8 in ID, 3/16 in OD
Catalog No. 502365931
Encompass_Preferred
PharMed BPT tubing is a biocompatible peristaltic pump tubing formulated for sensitive bioprocess and laboratory fluid transfer. This configuration has a 1/8 in inner diameter, 3/16 in outer diameter, and 1/32 in wall thickness, supplied in a standard 25 ft coil. The tubing provides extended flex life for peristaltic pumps, low extractables, and broad chemical compatibility for media processing, cell culture, and diagnostic workflows.
  • Biocompatible fluid contact surface, meets USP Class VI and ISO 10993-5 testing.
  • Formulated for peristaltic pumping with extended flex life and reduced downtime.
  • Temperature resistant from -59 °C to 135 °C for autoclave and hot-fluid applications.
  • Good chemical resistance to acids, alkalis, and oxidizers; compatible with common cleaners.
  • Minimizes particulate and extractables for sensitive bioprocessing workflows.
  • Standard coil length 25 ft for convenient installation and replacement.
Catalog No. 50-236-5931 Supplier Accuflow Systems Supplier No. AY242006
PharMed BPT tubing is a biocompatible peristaltic pump tubing formulated for sensitive bioprocess and laboratory fluid transfer. This configuration has a 1/8 in inner diameter, 3/16 in outer diameter, and 1/32 in wall thickness, supplied in a standard 25 ft coil. The tubing provides extended flex life for peristaltic pumps, low extractables, and broad chemical compatibility for media processing, cell culture, and diagnostic workflows.
  • Biocompatible fluid contact surface, meets USP Class VI and ISO 10993-5 testing.
  • Formulated for peristaltic pumping with extended flex life and reduced downtime.
  • Temperature resistant from -59 °C to 135 °C for autoclave and hot-fluid applications.
  • Good chemical resistance to acids, alkalis, and oxidizers; compatible with common cleaners.
  • Minimizes particulate and extractables for sensitive bioprocessing workflows.
  • Standard coil length 25 ft for convenient installation and replacement.