Learn More
REPLIGEN CORPORATION PHARMED TEE SAMPLE 1/8X1/4
SDP
Catalog No. NC1634953
Encompass
FLEXREADY™ Pharmed Tee Sample Line, 1/8" ID x1/4" OD PharMed BPT, NIP, plugs, Single Pouch,Gamma Irradiated at 25 - 40 kGy., Rev A, Ea.
Catalog No. NC1634953 Supplier REPLIGEN CORPORATION Supplier No. FXK00010445
FLEXREADY™ Pharmed Tee Sample Line, 1/8" ID x1/4" OD PharMed BPT, NIP, plugs, Single Pouch,Gamma Irradiated at 25 - 40 kGy., Rev A, Ea.