Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, PharMed BPT, 1/4"" ID x 3/8"" OD; 25 ft
Catalog No. NC9844461
Encompass
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165¿F (-46 to 73¿C). Can be sterilized with ethylene oxide or autoclave.
Catalog No. NC9844461 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9688010
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165¿F (-46 to 73¿C). Can be sterilized with ethylene oxide or autoclave.