Learn More
BD Pharmingen™ Indo-1 AM Dye
Catalog No. BDB565879 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
BDB565879 500 μg
1 options
Catalog No. BDB565879 Supplier BD Supplier No. 565879
Add to Cart
Add to Cart
Cell-permeant dye that can be used to measure intracellular ionized calcium concentration, [Ca2+]i. Indo-1 AM is a ratiometric dye: with UV excitation, the ratio of emission around 410nm to the emission around 485nm can be used to measure intracellular ionized calcium concentration.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Flow Cytometry
Quantity 500 μg