Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ MonoBlock™ Leukocyte Staining Buffer

Catalog No. BDB570004 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570004 25 Tests
BDB570003 100 Tests
BDB570002 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BDB570004 Supplier BD Supplier No. 570004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BD Pharmingen™ MonoBlock™ Leukocyte Staining Buffer reduces this nonspecific background staining, while not impacting specific surface staining or cell viability.

Some cyanine-type dyes (PE-Cy5, PerCP-Cy5.5, PE-Cy7, APC-Cy7, APC-H7, and others) and non-cyanine tandem dyes (eg, PE-CF594) are known to bind to human and mouse monocytes, macrophages, and other leukocyte subsets, though the exact mechanism is not clear. BD Pharmingen™ MonoBlock™ Leukocyte Staining Buffer reduces this nonspecific background staining, while not impacting specific surface staining or cell viability. This buffer is added directly to leukocytes or added to fluorescent reagent cocktails prior to staining leukocytes. This reagent may be complemented by others including i) BD Horizon™ Brilliant Stain Buffer, which mitigates dye-dye interactions amongst BD Horizon™ Brilliant fluorochromes, and ii) BD Fc Block™ reagents. The latter can reduce potential nonspecific staining caused by fluorescent antibodies that bind to Fc receptors expressed by human and mouse cells. For optimal and reproducible surface staining results, BD Pharmingen™ MonoBlock™ Leukocyte Staining Buffer should be used with cyanine-like and other tandem dye-conjugated reagents that can potentially stain cells nonspecifically. Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless to Yellow
Format Aqueous Liquid
For Use With (Application) Flow Cytometry
For Use With (Equipment) Flow Cytometer
Quantity 25 Tests
Product Type Staining Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.