Learn More
MSC Phase II Electronic Thickness Gage, 0.025 to 23 in, 0.01 mm Resolution
Catalog No. 5000528178
Encompass
This electronic thickness gage offers measurement capabilities from 0.025-23 in in Pulse Echo Mode and 2-23/64 in in Echo-Echo Mode. It provides a high resolution of 0.01 mm and an accuracy of up to 0.015 mm.
  • Dynamic functionality
  • Simple calibration process
  • Thru-coating measurement capability
  • Adjustable sound velocity
  • Multi-functional design
  • Includes certificate of calibration
  • NIST certified
  • Batteries included
  • Thumb button operating mechanism
  • Flat anvil tip shape
Catalog No. 50-005-28178 Supplier MSC Supplier No. 52845781
This electronic thickness gage offers measurement capabilities from 0.025-23 in in Pulse Echo Mode and 2-23/64 in in Echo-Echo Mode. It provides a high resolution of 0.01 mm and an accuracy of up to 0.015 mm.
  • Dynamic functionality
  • Simple calibration process
  • Thru-coating measurement capability
  • Adjustable sound velocity
  • Multi-functional design
  • Includes certificate of calibration
  • NIST certified
  • Batteries included
  • Thumb button operating mechanism
  • Flat anvil tip shape