Learn More
MSC Phase II Hardness Calibration Block, Rockwell C, Steel, 62 HRC
Catalog No. 5000529866
Encompass
This hardness calibration test block is designed for precise measurements on the Rockwell C scale. Made from durable steel, it features a C62 hardness for high range applications, ensuring accurate calibration of hardness testing equipment.
  • Test block type
  • Rockwell C scale type
  • Maximum hardness of 62 HRC
  • Steel material
  • High hardness range
  • Made in the USA
Catalog No. 50-005-29866 Supplier MSC Supplier No. 00444786
This hardness calibration test block is designed for precise measurements on the Rockwell C scale. Made from durable steel, it features a C62 hardness for high range applications, ensuring accurate calibration of hardness testing equipment.
  • Test block type
  • Rockwell C scale type
  • Maximum hardness of 62 HRC
  • Steel material
  • High hardness range
  • Made in the USA