Learn More
MSC Phase II Portable Electronic Hardness Tester, 200-960 HL, Rockwell, Brinell, Vickers
Catalog No. 5000527485
Encompass
This portable electronic hardness tester measures hardness within a range of 200 HL to 960 HL. It is capable of measuring across multiple scales, including Brinell, Rockwell B, Rockwell C, Shore, and Vickers, with an accuracy of up to 0.5%. Designed for robust applications, it can measure the hardness of heavy steel masses at least 1 inch thick, including round parts.
  • Measures hardness from 200 HL to 960 HL.
  • Compatible with Brinell, Rockwell B/C, Shore, and Vickers scales.
  • Provides accuracy up to 0.5%.
  • Features a 3 mm ball indenter.
  • Powered by Alkaline AA batteries.
  • Measures HRC from 20-70.
  • Includes base instrument, impact device, cleaning brush, calibrated test block, custom carry case, and operation manual.
Catalog No. 50-005-27485 Supplier MSC Supplier No. 74389982
This portable electronic hardness tester measures hardness within a range of 200 HL to 960 HL. It is capable of measuring across multiple scales, including Brinell, Rockwell B, Rockwell C, Shore, and Vickers, with an accuracy of up to 0.5%. Designed for robust applications, it can measure the hardness of heavy steel masses at least 1 inch thick, including round parts.
  • Measures hardness from 200 HL to 960 HL.
  • Compatible with Brinell, Rockwell B/C, Shore, and Vickers scales.
  • Provides accuracy up to 0.5%.
  • Features a 3 mm ball indenter.
  • Powered by Alkaline AA batteries.
  • Measures HRC from 20-70.
  • Includes base instrument, impact device, cleaning brush, calibrated test block, custom carry case, and operation manual.