Learn More
Sensorex pHASE pH Sensor, Glass,BNC
Catalog No. 501897526
Encompass_Preferred
The research grade pH3400 is a highly versatile combination pH electrode, specially designed to respond quickly and accurately to rapid temperature changes.
Catalog No. 50-189-7526 Supplier Sensorex Supplier No. SG1041CD
The research grade pH3400 is a highly versatile combination pH electrode, specially designed to respond quickly and accurately to rapid temperature changes.