Learn More
Sensorex PHASE RENEW PH ELECTRODE
Catalog No. NC0155074
Encompass_Preferred
pHASE Rapid renew pH electrode, plastic body, removable bulb guard, 1meter cable BNC connector
Catalog No. NC0155074 Supplier Sensorex Supplier No. S1021CD/BNC
pHASE Rapid renew pH electrode, plastic body, removable bulb guard, 1meter cable BNC connector