Learn More
Taylor Wharton Model 82 Phase Separator, 1 1/4″ x 1″ OD, 3/8″ FNPT
Catalog No. NC9309108
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. PHASE SEPARATOR 1IN
Catalog No. NC9309108 Supplier Taylor Wharton Supplier No. 11938C82
This item has a minimum order quantity of 2 per supplier requirements. PHASE SEPARATOR 1IN