Learn More
Aviva Systems Biology Corp PHAX Antibody - C-terminal region (ARP65475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHAX. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human PHAX. Alternate names: FLJ13193, RNUXAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-505