Learn More
Aviva Systems Biology Corp PHB2 Antibody - C-terminal region (ARP48316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PHB2. Alternate names: BAP, BCAP37, Bap37, MGC117268, PNAS-141, REA, p22ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59795