Learn More
Aviva Systems Biology Corp Phb2 Antibody - middle region (ARP61279_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61279_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Phb2. It was validated on Western Blot. . Alternate names: AU044498, BAP, Bap37, Bcap37, REAApplicationWB, IP. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-324