Learn More
Lonza Walkersville Human Peripheral Blood Mononuclear Cells (hPBMC), Cryopreserved, 50 million
Catalog No. NC1948958
Encompass
Lonza is your ideal partner when you are looking to buy PBMC. Our Peripheral Blood Mononuclear Cells (PBMCs) are isolated from peripheral blood by apheresis and density gradient separation. They are guaranteed to contain ≥50 million viable cells per ampoule. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis (CoA) is provided for each cell lot purchasedApplications:ELISPOTProliferation assaysImmunogenicity assaysCell therapy process developmentHumanized miceBiomarker discoveryContent:1 x Cryopreserved ampule of Human Peripheral Blood Mononuclear Cells (hPBMC) containing ≥ 50 million cells
This item is not returnable. View return policy
Catalog No. NC1948958 Supplier Lonza Walkersville Supplier No. CC2702
Lonza is your ideal partner when you are looking to buy PBMC. Our Peripheral Blood Mononuclear Cells (PBMCs) are isolated from peripheral blood by apheresis and density gradient separation. They are guaranteed to contain ≥50 million viable cells per ampoule. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis (CoA) is provided for each cell lot purchasedApplications:ELISPOTProliferation assaysImmunogenicity assaysCell therapy process developmentHumanized miceBiomarker discoveryContent:1 x Cryopreserved ampule of Human Peripheral Blood Mononuclear Cells (hPBMC) containing ≥ 50 million cells