Learn More
Enzo Life Sciences PHC-SKKKK 3 TFA (1 mg)
SDP
Catalog No. NC1735356
Encompass_Preferred
Negative control for Pam3Cys-Ser-(Lys)4 . trihydrochloride. Does not activate TLR2. Can be used as negative control for Pam3Cys-Ser-(Lys)4 . trihydrochloride (Prod. No. ALX-165-066). Alternative Name: PHCSK4 . 3 TFA, N-palmitoyl-S-(1,2-bishexadecyloxy- carbonyl)-ethyl-(R)-cysteinyl-seryl-(lysyl)3-lysine. Formula: C82H158N10O13S . 3 CF3COOH. MW: 1524.3. 342.1. Formulation: Lyophilized. Appearance: Colorless powder. Solubility: Soluble in pyrogen-free deionized water, DMSO or aqueous buffers at pH <7.5. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. NC1735356 Supplier Enzo Life Sciences Supplier No. ALX165071M001
Negative control for Pam3Cys-Ser-(Lys)4 . trihydrochloride. Does not activate TLR2. Can be used as negative control for Pam3Cys-Ser-(Lys)4 . trihydrochloride (Prod. No. ALX-165-066). Alternative Name: PHCSK4 . 3 TFA, N-palmitoyl-S-(1,2-bishexadecyloxy- carbonyl)-ethyl-(R)-cysteinyl-seryl-(lysyl)3-lysine. Formula: C82H158N10O13S . 3 CF3COOH. MW: 1524.3. 342.1. Formulation: Lyophilized. Appearance: Colorless powder. Solubility: Soluble in pyrogen-free deionized water, DMSO or aqueous buffers at pH <7.5. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C.