Learn More
Hach Company Intellical™ PHC108 Laboratory Puncture Stainless Steel pH Electrode, 1 m Cable
Catalog No. 501998075 Shop All Hach Company Products
Encompass
Intellical™ PHC108 is a digital combination puncture pH electrode with non-refillable colored electrolyte (polymer) with a ceramic pin plus an open reference junctions, and built-in temperature sensor. The PHC108 has an easy to clean stainless steel body perfect for use in the food industries. Thanks to its spear-shaped glass sensor the PHC108 is specifically designed to measure pH in semi-solid samples. The PHC108 is not suitable for use with organic solvents, low ionic strength or high alkaline sample types.
Catalog No. 50-199-8075 Supplier Hach Company Supplier No. PHC10801
Intellical™ PHC108 is a digital combination puncture pH electrode with non-refillable colored electrolyte (polymer) with a ceramic pin plus an open reference junctions, and built-in temperature sensor. The PHC108 has an easy to clean stainless steel body perfect for use in the food industries. Thanks to its spear-shaped glass sensor the PHC108 is specifically designed to measure pH in semi-solid samples. The PHC108 is not suitable for use with organic solvents, low ionic strength or high alkaline sample types.