Learn More
Aviva Systems Biology Corp PHC2 Antibody - N-terminal region (ARP51251_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51251_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHC2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PHC2. Alternate names: EDR2, HPH2, MGC163502, PH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-568