Learn More
ABclonal Technology PHC2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14610500UL
Encompass_Preferred
In Drosophila melanogaster, the Polycomb group (PcG) of genes are part of a cellular memory system that is responsible for the stable inheritance of gene activity. PcG proteins form a large multimeric, chromatin-associated protein complex. The protein encoded by this gene has homology to the Drosophila PcG protein polyhomeotic (Ph) and is known to heterodimerize with EDR1 and colocalize with BMI1 in interphase nuclei of human cells. The specific function in human cells has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-06395