Learn More
Hach Company Intellical™ PHC705 Laboratory General Purpose RedRod Refillable Glass pH Electrode, 1 m Cable
Catalog No. 501998082 Shop All Hach Company Products
Encompass
Intellical™ PHC705 is a digital combination pH electrode with built-in temperature sensor. The PHC705 is a pH refillable electrode with ceramic porous pin reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. This high performance laboratory glass pH electrode is ideal for measuring pH in a variety of laboratory applications for aqueous samples. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC705 is not suitable for use with organic solvents, emulsions, samples with high solids content or high alkaline sample types.
Catalog No. 50-199-8082 Supplier Hach Company Supplier No. PHC70501
Intellical™ PHC705 is a digital combination pH electrode with built-in temperature sensor. The PHC705 is a pH refillable electrode with ceramic porous pin reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. This high performance laboratory glass pH electrode is ideal for measuring pH in a variety of laboratory applications for aqueous samples. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC705 is not suitable for use with organic solvents, emulsions, samples with high solids content or high alkaline sample types.