Learn More
Hach Company Intellical™ PHC705 Laboratory High Alkalinity Range RedRod Refillable Glass pH Electrode, 1 m Cable
Catalog No. 501998083 Shop All Hach Company Products
Encompass
Intellical™ PHC705A is a digital combination pH electrode with built-in temperature sensor. The PHC705A is a pH refillable electrode with ceramic porous pin reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its special pH glass bulb composition this high performance laboratory glass pH electrode is particularly suited for measuring pH in alkaline aqueous samples for pH above 12. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC705 is not suitable for use with organic solvents, emulsions or sample types with high solids content.
Catalog No. 50-199-8083 Supplier Hach Company Supplier No. PHC705A01
Intellical™ PHC705A is a digital combination pH electrode with built-in temperature sensor. The PHC705A is a pH refillable electrode with ceramic porous pin reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its special pH glass bulb composition this high performance laboratory glass pH electrode is particularly suited for measuring pH in alkaline aqueous samples for pH above 12. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC705 is not suitable for use with organic solvents, emulsions or sample types with high solids content.