Learn More
Hach Company Intellical™ PHC725 Laboratory Low Ionic Strength Media RedRod Refillable Glass pH Electrode, 1 m Cable
Catalog No. 501998084 Shop All Hach Company Products
Encompass
Intellical™ PHC725 is a digital combination pH electrode with built-in temperature sensor. The PHC725 is a pH refillable electrode with annular ceramic porous reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its annular ring reference junction this high performance laboratory glass pH electrode is ideal for measuring pH in Low Ionic Strength aqueous solutions. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC725 is not suitable for use with organic solvents, emulsions, samples with high solids content or high alkaline sample types.
Catalog No. 50-199-8084 Supplier Hach Company Supplier No. PHC72501
Intellical™ PHC725 is a digital combination pH electrode with built-in temperature sensor. The PHC725 is a pH refillable electrode with annular ceramic porous reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its annular ring reference junction this high performance laboratory glass pH electrode is ideal for measuring pH in Low Ionic Strength aqueous solutions. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC725 is not suitable for use with organic solvents, emulsions, samples with high solids content or high alkaline sample types.