Learn More
Hach Company Intellical™ PHC729 Laboratory Surface Measurements RedRod Refillable Glass pH Electrode, 1 m Cable
Catalog No. 501998085 Shop All Hach Company Products
Encompass
Intellical™ PHC729 is a digital combination pH electrode with built-in temperature sensor. The PHC729 is a pH refillable flat sensor electrode with annular ceramic porous reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to the combination of the flat pH sensing element and the annular ring reference junction this high performance laboratory glass pH electrode is ideal for measuring pH on flat surfaces of various samples or where there is a minimal sample depth. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC729 is not suitable for use with organic solvents, emulsions, samples with high solids content orhigh alkaline sample types.
Catalog No. 50-199-8085 Supplier Hach Company Supplier No. PHC72901
Intellical™ PHC729 is a digital combination pH electrode with built-in temperature sensor. The PHC729 is a pH refillable flat sensor electrode with annular ceramic porous reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to the combination of the flat pH sensing element and the annular ring reference junction this high performance laboratory glass pH electrode is ideal for measuring pH on flat surfaces of various samples or where there is a minimal sample depth. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC729 is not suitable for use with organic solvents, emulsions, samples with high solids content orhigh alkaline sample types.