Learn More
Hach Company Intellical™ PHC735 Laboratory Dirty Media RedRod Refillable Glass pH Electrode, 1 m Cable
Catalog No. 501998086 Shop All Hach Company Products
Encompass
Intellical™ PHC735 is a digital combination pH electrode with built-in temperature sensor. The PHC735 is a pH refillable electrode with a high flow annular ceramic porous reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its high flow annular ring reference junction this high performance laboratory glass pH electrode is ideal for measuring pH in “dirty” samples, including wastewater, food and beverage applications. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC735 is not suitable for use with organic solvents or high alkaline sample types.
Catalog No. 50-199-8086 Supplier Hach Company Supplier No. PHC73501
Intellical™ PHC735 is a digital combination pH electrode with built-in temperature sensor. The PHC735 is a pH refillable electrode with a high flow annular ceramic porous reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its high flow annular ring reference junction this high performance laboratory glass pH electrode is ideal for measuring pH in “dirty” samples, including wastewater, food and beverage applications. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC735 is not suitable for use with organic solvents or high alkaline sample types.