Learn More
Hach Company Intellical™ PHC745 Laboratory Clogging Media RedRod Refillable Glass pH Electrode, 1 m Cable
Catalog No. 501998087 Shop All Hach Company Products
Encompass
Intellical™ PHC745 is a digital combination pH electrode with built-in temperature sensor. The PHC745 is a pH refillable electrode with a reverse sleeve reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its easy to clean and high flow reverse sleeve reference junction this high performance laboratory glass pH electrode is ideal for measuring pH in samples prone to clogging traditional reference junctions such as wastewater influents, emulsions and viscous liquids. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC745 is not suitable for use with organic solvents or high alkaline sample types.
Catalog No. 50-199-8087 Supplier Hach Company Supplier No. PHC74501
Intellical™ PHC745 is a digital combination pH electrode with built-in temperature sensor. The PHC745 is a pH refillable electrode with a reverse sleeve reference junction embedding the RedRod technology to deliver exceptionally fast, stable, and accurate response times for accurate measurements within a wide temperature range. Thanks to its easy to clean and high flow reverse sleeve reference junction this high performance laboratory glass pH electrode is ideal for measuring pH in samples prone to clogging traditional reference junctions such as wastewater influents, emulsions and viscous liquids. A 59-mL bottle of saturated KCl Electrode Filling Solution is included with the probe. The PHC745 is not suitable for use with organic solvents or high alkaline sample types.