Learn More
Hach Company Intellical™ PHC805 Laboratory General Purpose Refillable Glass pH Electrode, 1 m Cable
Catalog No. 501998088 Shop All Hach Company Products
Encompass
Intellical™ PHC805 is a digital combination pH electrode with built-in temperature sensor. The PHC805 is a pH refillable electrode with ceramic porous pin reference junction. This laboratory glass pH electrode is ideal for measuring pH in general purposes laboratory applications for aqueous samples. A 50-mL bottle of KCl 3M Electrode Filling Solution is included with the probe. The PHC805 is not suitable for use with organic solvents, low ionic strength, wastewater, high solids or high alkaline sample types.
Catalog No. 50-199-8088 Supplier Hach Company Supplier No. PHC80501
Intellical™ PHC805 is a digital combination pH electrode with built-in temperature sensor. The PHC805 is a pH refillable electrode with ceramic porous pin reference junction. This laboratory glass pH electrode is ideal for measuring pH in general purposes laboratory applications for aqueous samples. A 50-mL bottle of KCl 3M Electrode Filling Solution is included with the probe. The PHC805 is not suitable for use with organic solvents, low ionic strength, wastewater, high solids or high alkaline sample types.