Learn More
Abcam PHCCC, allosteric potentiator at mGlu4, 50MG
Catalog No. 502707903
Encompass_Preferred
MW 294.3 Da, Purity >98%. Selective allosteric potentiator of mGlu4. Anti-Parkinson and anxiolytic effects *in vivo*. Also antagonist at Group I metabotropic receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7903 Supplier Abcam Supplier No. AB12004350MG
MW 294.3 Da, Purity >98%. Selective allosteric potentiator of mGlu4. Anti-Parkinson and anxiolytic effects *in vivo*. Also antagonist at Group I metabotropic receptors.
The product is subject to the following: Abcam Restricted Use Statement