Learn More
Strem, An Ascensus Company CAS# 1447963-71-4. 100MG. 2-Diphenylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl, min. 98% PhCPhos
SDP
Catalog No. 501963553
Encompass
CAS# 1447963-71-4. 100MG. 2-Diphenylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl, min. 98%. PhCPhos Molecular Formula: C28H29N2P. Molecular Weight: 424.52. Color/Form: tan solid. Strem# 15-1125. www.strem.com/catalog/v3/15-1125/
Catalog No. 50-196-3553 Supplier Strem, An Ascensus Company Supplier No. 151125100MG
CAS# 1447963-71-4. 100MG. 2-Diphenylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl, min. 98%. PhCPhos Molecular Formula: C28H29N2P. Molecular Weight: 424.52. Color/Form: tan solid. Strem# 15-1125. www.strem.com/catalog/v3/15-1125/