Learn More
Medchemexpress LLC PHD-1-IN-1 | 2009343-14-8 | 99.2% | 220.23 | 50 MG
SDP
Catalog No. 5000349697
Encompass_Preferred
PHD-1-IN-1 is an orally active and potent HIF prolylhydroxylase domain-1 (PHD-1) inhibitor with an IC50 of 0.034 μM. It has a unique monodentate binding interaction with the active site Fe2+ ion and induces the formation of an "Arg367-out" pocket.
  • Orally active and potent HIF prolylhydroxylase domain-1 (PHD-1) inhibitor
  • IC50 of 0.034 μM (PHD-1)
  • Unique monodentate binding interaction with the active site Fe2+ ion
  • Induces the formation of an "Arg367-out" pocket
Catalog No. 50-003-49697 Supplier Medchemexpress LLC Supplier No. HY13630050MG
PHD-1-IN-1 is an orally active and potent HIF prolylhydroxylase domain-1 (PHD-1) inhibitor with an IC50 of 0.034 μM. It has a unique monodentate binding interaction with the active site Fe2+ ion and induces the formation of an "Arg367-out" pocket.
  • Orally active and potent HIF prolylhydroxylase domain-1 (PHD-1) inhibitor
  • IC50 of 0.034 μM (PHD-1)
  • Unique monodentate binding interaction with the active site Fe2+ ion
  • Induces the formation of an "Arg367-out" pocket