Learn More
Medchemexpress LLC Phd-1-in-1 | 2009343-14-8 | 99.2% | 220.23 | C13H8N4 | 5 MG
SDP
Catalog No. 5000222811
Encompass_Preferred
PHD-1-IN-1 is a potent, orally active small-molecule inhibitor of hypoxia-inducible factor prolyl-hydroxylase domain-1 (PHD-1) for preclinical research into HIF regulation and hypoxia signaling. It binds the active-site Fe2+ with a unique monodentate interaction and induces an Arg367-out pocket. Supplied as a white to off-white solid with high purity for in vitro and in vivo studies.
  • Potent PHD-1 inhibition (IC50 0.034 μM).
  • Unique monodentate binding to the active-site iron that alters pocket conformation.
  • High purity suitable for research and assay development.
  • Soluble in DMSO; manufacturer provides in vivo formulation protocols.
  • Stable powder storage conditions for long-term preservation.
Catalog No. 50-002-22811 Supplier Medchemexpress LLC Supplier No. HY1363005MG
PHD-1-IN-1 is a potent, orally active small-molecule inhibitor of hypoxia-inducible factor prolyl-hydroxylase domain-1 (PHD-1) for preclinical research into HIF regulation and hypoxia signaling. It binds the active-site Fe2+ with a unique monodentate interaction and induces an Arg367-out pocket. Supplied as a white to off-white solid with high purity for in vitro and in vivo studies.
  • Potent PHD-1 inhibition (IC50 0.034 μM).
  • Unique monodentate binding to the active-site iron that alters pocket conformation.
  • High purity suitable for research and assay development.
  • Soluble in DMSO; manufacturer provides in vivo formulation protocols.
  • Stable powder storage conditions for long-term preservation.