Learn More
Hach Company Hach Online Process ORP Sensor - General Purpose Analog ORP Sensor
Catalog No. 501998089 Shop All Hach Company Products
Encompass
General Purpose Online Process ORP Sensor - Platinum ORP Electrode, PEEK Housing, Convertible Mount, 4.5m Cable, Analog Cable Connection
Catalog No. 50-199-8089 Supplier Hach Company Supplier No. RD1P5
General Purpose Online Process ORP Sensor - Platinum ORP Electrode, PEEK Housing, Convertible Mount, 4.5m Cable, Analog Cable Connection