Learn More
Cayman Chemical Phe-Arg-βNA (hydrochloride), Purity: ≥98%, CAS: 100929-99-5, 250 mg
SDP
Catalog No. 502988827
Encompass_Preferred
A broad-spectrum bacterial efflux pump inhibitor; potentiates the activity of levofloxacin against P. aeruginosa strains overexpressing the MexAB-OprM, MexCD-OprJ, or MexEF-OprN multidrug resistance efflux pumps (EC50s = 10 µg/ml for all); increases the susceptibility of quinolone-resistant E. coli isolates to nalidixic acid
Catalog No. 50-298-8827 Supplier Cayman Chemical Supplier No. 29511250 MG
A broad-spectrum bacterial efflux pump inhibitor; potentiates the activity of levofloxacin against P. aeruginosa strains overexpressing the MexAB-OprM, MexCD-OprJ, or MexEF-OprN multidrug resistance efflux pumps (EC50s = 10 µg/ml for all); increases the susceptibility of quinolone-resistant E. coli isolates to nalidixic acid