Learn More
Medchemexpress LLC Phe-Arg-β-naphthylamide dihydrochloride | 100929-99-5 | MFCD00058046 | 99.9% | 519.47 g/mol | C25H32Cl2N6O2 | 10 MG
SDP
Catalog No. 5000245354
Encompass_Preferred
PAβN dihydrochloride (Phe-Arg-β-naphthylamide dihydrochloride) is a research reagent that inhibits bacterial multidrug efflux pumps and is used to probe efflux-mediated antibiotic resistance and membrane permeability. Supplied as the dihydrochloride salt for improved solubility, it is intended for use in microbiology and biochemical assays focused on Gram-negative transporters.
  • Efflux pump inhibitor active against Mex-family transporters.
  • High purity (≈99.9%).
  • Molecular weight 519.47 g/mol; CAS 100929-99-5.
  • Off-white to light yellow solid supplied as the dihydrochloride salt.
  • Suitable for bacterial efflux inhibition and permeability assays.
  • Typical quantity 10 MG vial.
Catalog No. 50-002-45354 Supplier Medchemexpress LLC Supplier No. HY101444A10MG
PAβN dihydrochloride (Phe-Arg-β-naphthylamide dihydrochloride) is a research reagent that inhibits bacterial multidrug efflux pumps and is used to probe efflux-mediated antibiotic resistance and membrane permeability. Supplied as the dihydrochloride salt for improved solubility, it is intended for use in microbiology and biochemical assays focused on Gram-negative transporters.
  • Efflux pump inhibitor active against Mex-family transporters.
  • High purity (≈99.9%).
  • Molecular weight 519.47 g/mol; CAS 100929-99-5.
  • Off-white to light yellow solid supplied as the dihydrochloride salt.
  • Suitable for bacterial efflux inhibition and permeability assays.
  • Typical quantity 10 MG vial.