Learn More
Medchemexpress LLC Phe-Arg-β-naphthylamide dihydrochloride | 100929-99-5 | MFCD00058046 | 99.9% | 519.47 g/mol | C25H32Cl2N6O2 | 25 MG
SDP
Catalog No. 5000245771
Encompass_Preferred
PAβN dihydrochloride is a small-molecule bacterial efflux pump inhibitor used in microbiology research to block multidrug resistance transporters and sensitize bacteria to antibiotics. It is supplied as an off-white to light-yellow solid with high listed purity and is typically used in in vitro assays to study antibiotic susceptibility and resistance mechanisms.
  • Inhibits gram-negative bacterial efflux pumps, reducing antibiotic minimum inhibitory concentrations.
  • Useful for in vitro studies of resistance mechanisms and susceptibility assays.
  • High purity suitable for research-grade experiments.
  • Solid form simplifies storage and handling in laboratory settings.
  • Provided in research-scale packaging appropriate for experimental use.
Catalog No. 50-002-45771 Supplier Medchemexpress LLC Supplier No. HY101444A25MG
PAβN dihydrochloride is a small-molecule bacterial efflux pump inhibitor used in microbiology research to block multidrug resistance transporters and sensitize bacteria to antibiotics. It is supplied as an off-white to light-yellow solid with high listed purity and is typically used in in vitro assays to study antibiotic susceptibility and resistance mechanisms.
  • Inhibits gram-negative bacterial efflux pumps, reducing antibiotic minimum inhibitory concentrations.
  • Useful for in vitro studies of resistance mechanisms and susceptibility assays.
  • High purity suitable for research-grade experiments.
  • Solid form simplifies storage and handling in laboratory settings.
  • Provided in research-scale packaging appropriate for experimental use.