Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™

Catalog No. ICN15612883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10mg
25mg
50mg
250mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15612883 250mg
ICN15612810 10mg
ICN15612825 25mg
ICN15612850 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15612883 Supplier MP Biomedicals, Inc Supplier No. 0215612883
Add to Cart
Add to Cart

Specifications

Purity >99%
Assay 0.854
CAS 100929-99-5
Color White
Content And Storage Store at 0°C.
Format Powder
Molecular Weight (g/mol) 519.471
Product Type Phe-Arg-β-naphthylamide dihydrochloride
Quantity 250mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.