Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phe-Lys(Fmoc)-PAB | 2149584-03-0 | MFCD28899327 | >99.0% | 620.74 g·mol⁻¹ | C37H40N4O5 | 25 MG
SDP

Catalog No. 5000202464
Encompass_Preferred

Phe-Lys(Fmoc)-PAB is a cathepsin-cleavable antibody-drug conjugate (ADC) linker used in the synthesis of ADCs. Supplied as a research reagent, it is intended for laboratory research use only.

  • Cathepsin-cleavable ADC linker for ADC synthesis.
  • Intended for research use only.
  • Available in multiple package sizes (5 mg-500 mg).
  • Purity >99.0%.
  • CAS number 2149584-03-0.
  • Molecular formula C37H40N4O5.
  • Molecular weight 620.74 g·mol⁻¹.

Catalog No. 50-002-02464 Supplier Medchemexpress LLC Supplier No. HY12936225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phe-Lys(Fmoc)-PAB is a cathepsin-cleavable antibody-drug conjugate (ADC) linker used in the synthesis of ADCs. Supplied as a research reagent, it is intended for laboratory research use only.

  • Cathepsin-cleavable ADC linker for ADC synthesis.
  • Intended for research use only.
  • Available in multiple package sizes (5 mg-500 mg).
  • Purity >99.0%.
  • CAS number 2149584-03-0.
  • Molecular formula C37H40N4O5.
  • Molecular weight 620.74 g·mol⁻¹.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.