Learn More
Medchemexpress LLC Phe-Lys(Trt)-PAB | 1116085-99-4 | 96.2% | 640.81 | C41H44N4O3 | 5 MG
SDP
Catalog No. 5000203131
Encompass_Preferred
Phe-Lys(Trt)-PAB is a cathepsin-cleavable peptide-PAB linker used in antibody-drug conjugate (ADC) research. It couples cytotoxic payloads to antibodies via a protease-sensitive spacer that remains stable in circulation and is cleaved by lysosomal cathepsins after internalization to release the payload. Supplied as a 5 mg solid for research use; store as powder at -20°C.
  • Cathepsin-cleavable peptide spacer.
  • Enables controlled lysosomal release of payloads.
  • Stable under physiological conditions.
  • High purity suitable for research applications.
  • Powder storage at -20°C extends shelf life.
Catalog No. 50-002-03131 Supplier Medchemexpress LLC Supplier No. HY1293495MG
Phe-Lys(Trt)-PAB is a cathepsin-cleavable peptide-PAB linker used in antibody-drug conjugate (ADC) research. It couples cytotoxic payloads to antibodies via a protease-sensitive spacer that remains stable in circulation and is cleaved by lysosomal cathepsins after internalization to release the payload. Supplied as a 5 mg solid for research use; store as powder at -20°C.
  • Cathepsin-cleavable peptide spacer.
  • Enables controlled lysosomal release of payloads.
  • Stable under physiological conditions.
  • High purity suitable for research applications.
  • Powder storage at -20°C extends shelf life.