Learn More
Medchemexpress LLC HY-P0249A 5mg Medchemexpress, Phe-Met-Arg-Phe amide trifluoroacetate CAS:159237-99-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0249A5MG
Encompass_Preferred
Medchemexpress, HY-P0249A 5mg Phe-Met-Arg-Phe amide trifluoroacetate CAS:159237-99-7 Phe-Met-Arg-Phe amide trifluoroacetate is an activator of K + current, with ED 50 of 23 nM in the peptidergic caudodorsal neurons. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4078