Learn More
Carbosynth LLC. Phe4,9 [Ring D5]-Hepcidin (Rat), 0.1mg, Disulfide bonds: Cys7-Cys23, Cys10-Cys13, Cys11-Cys19, and Cys14-Cys22, MW: (M.W. 2722.37), Molecular formula: C111H161D10N29O34S8
SDP
Supplier:  Carbosynth LLC. PLP3780PI0.1MG
Encompass_Preferred
Synonyms: Asp-Thr-Asn-Phe(D5)-Pro-Ile-Cys-Leu-Phe(D5)-Cys-Cys-Lys-Cys-Cys-Lys-Asn-Ser-Ser-Cys-Gly-Leu-Cys-Cys-Ile-Thr, Application: Deuterium Stable Isotope-Labeled Hepcidin for Internal Standard, Storage Temperature: -20°C
Catalog No. 50-168-6610