Learn More
Cole-Parmer Oakton pH Electrode, Refillable, Epoxy, Double-Junction, 12 mm OD x 110 mm L; 3-ft BNC Cable
Catalog No. 501961372 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1372 Supplier Cole-Parmer Supplier No. 3580500
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.